siRNA Details
| Gene Symbol: Fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109606.1 | NP_001103076.1 | 9, 10 | 1075 | ||
| XM_006234656.2 | XP_006234718.1 | 9, 10 | 1100 | |||
| XM_006234657.2 | XP_006234719.1 | 9, 10 | 1100 | |||
| GenBank mRNA | FQ210238.1 | - | 2, 3 | 218 | ||

Fbxo3
Genomic Map