siRNA Details
| Gene Symbol: Wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168641.1 | NP_001162112.1 | 5, 6 | 636 | ||
| XM_006233028.2 | XP_006233090.1 | 4, 5 | 2499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wars2| Gene Symbol: Wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168641.1 | NP_001162112.1 | 5, 6 | 636 | ||
| XM_006233028.2 | XP_006233090.1 | 4, 5 | 2499 | |||