siRNA Details
| Gene Symbol: Zfp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168642.1 | NP_001162113.1 | 6 | 661 | ||
| XM_006228106.2 | XP_006228168.1 | 6 | 704 | |||
| XM_008758775.1 | XP_008756997.1 | 6 | 720 | |||
| XM_008758776.1 | XP_008756998.1 | 4 | 345 | |||

Zfp40
Genomic Map