siRNA Details
| Gene Symbol: LOC100910998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768248.1 | XP_008766470.1 | 219 | |||
| Gene Symbol: LOC103693424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768186.1 | XP_008766408.1 | 268 | |||
| Gene Symbol: LOC103693451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768247.1 | XP_008766469.1 | 306 | |||
| Gene Symbol: Tmem92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768249.1 | XP_008766471.1 | 578 | |||

LOC100910998
Genomic Map