siRNA Details
| Gene Symbol: Ddx19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772534.1 | XP_008770756.1 | 7 | 625 | ||
| GenBank mRNA | EV774940.1 | - | 7 | 710 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx19b| Gene Symbol: Ddx19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772534.1 | XP_008770756.1 | 7 | 625 | ||
| GenBank mRNA | EV774940.1 | - | 7 | 710 | ||