siRNA Details
Gene Symbol: LOC363181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC082066.1 | - | 5, 6 | 1026 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC363181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC082066.1 | - | 5, 6 | 1026 |