siRNA Details
| Gene Symbol: Vom2r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228107.2 | XP_006228169.2 | 2 | 1157 | ||
| GenBank mRNA | AF053991.1 | - | 1 | 543 | ||
| Gene Symbol: LOC100910798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750426.3 | XP_003750474.2 | 2506 | |||
| Gene Symbol: Vom2r19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099492.1 | NP_001092962.1 | 6 | 2506 | ||
More Information
Set Membership: |
|

Vom2r21
Genomic Map