siRNA Details
| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772570.1 | XP_008770792.1 | 9 | 1484 | ||
| XM_008772571.1 | XP_008770793.1 | 9 | 1439 | |||
| XM_008772572.1 | XP_008770794.1 | 8 | 1350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlkl| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772570.1 | XP_008770792.1 | 9 | 1484 | ||
| XM_008772571.1 | XP_008770793.1 | 9 | 1439 | |||
| XM_008772572.1 | XP_008770794.1 | 8 | 1350 | |||