siRNA Details
| Gene Symbol: HS3ST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006040.2 | NP_006031.2 | 2 | 1720 | ||
| GenBank mRNA | AF105378.2 | - | 2 | 1717 | ||
| AL834283.1 | - | 2 | 967 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS3ST4| Gene Symbol: HS3ST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006040.2 | NP_006031.2 | 2 | 1720 | ||
| GenBank mRNA | AF105378.2 | - | 2 | 1717 | ||
| AL834283.1 | - | 2 | 967 | |||