siRNA Details
| Gene Symbol: LOC100911583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750446.2 | XP_003750494.2 | 2 | 739 | ||
| Gene Symbol: LOC690764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766033.1 | XP_008764255.1 | 655 | |||

LOC100911583
Genomic Map