siRNA Details
| Gene Symbol: HS3ST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006040.2 | NP_006031.2 | 2 | 1296 | ||
| GenBank mRNA | AF105378.2 | - | 2 | 1293 | ||
| AL834283.1 | - | 2 | 543 | |||
| EF063601.1 | - | 2 | 367 | |||
| EF063602.1 | - | 2 | 367 | |||
| EF063603.1 | - | 2 | 367 | |||
| EF063604.1 | - | 2 | 367 | |||
| EF063605.1 | - | 2 | 367 | |||
| EF063606.1 | - | 2 | 367 | |||
| EF063607.1 | - | 2 | 367 | |||
| EF063608.1 | - | 2 | 367 | |||

HS3ST4
Genomic Map