siRNA Details
| Gene Symbol: LOC690784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044299.1 | NP_001037764.1 | 2 | 393 | ||
| XM_008763425.1 | XP_008761647.1 | 1 | 1499 | |||
| XM_008763426.1 | XP_008761648.1 | 2 | 827 | |||
| XM_008763427.1 | XP_008761649.1 | 1 | 1499 | |||
| XM_008763428.1 | XP_008761650.1 | 1 | 1499 | |||
| XM_008763429.1 | XP_008761651.1 | 1 | 1499 | |||
| XM_008763430.1 | XP_008761652.1 | 1 | 1499 | |||
| GenBank mRNA | BC082012.1 | - | 2 | 393 | 7132059 | |

LOC690784
Genomic Map