siRNA Details
| Gene Symbol: Gngt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135767.1 | NP_001129239.1 | 4 | 341 | ||
| XM_006247208.2 | XP_006247270.1 | 4 | 452 | |||
| XM_006247209.1 | XP_006247271.1 | 4 | 250 | |||
| XM_006247210.1 | XP_006247272.1 | 4 | 250 | |||
| XM_006247211.2 | XP_006247273.1 | 3 | 500 | |||
| XM_008768115.1 | XP_008766337.1 | 3 | 397 | |||

Gngt2
Genomic Map