siRNA Details
| Gene Symbol: Aatk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168703.1 | NP_001162174.1 | 7 | 881 | ||
| XM_006247926.2 | XP_006247988.1 | 7 | 993 | |||
| XM_006247927.2 | XP_006247989.1 | 6 | 674 | |||
| XM_006247928.2 | XP_006247990.1 | 7 | 885 | |||
| XM_008768484.1 | XP_008766706.1 | 7 | 906 | |||

Aatk
Genomic Map