siRNA Details
| Gene Symbol: Aatk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168703.1 | NP_001162174.1 | 13 | 4140 | ||
| XM_006247926.2 | XP_006247988.1 | 13 | 4252 | |||
| XM_006247927.2 | XP_006247989.1 | 12 | 3933 | |||
| XM_006247928.2 | XP_006247990.1 | 13 | 4144 | |||
| XM_008768484.1 | XP_008766706.1 | 13 | 4165 | |||

Aatk
Genomic Map