siRNA Details
| Gene Symbol: Sh3bp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127581.1 | NP_001121053.1 | 6 | 1479 | ||
| XM_008767708.1 | XP_008765930.1 | 7 | 1596 | |||
| XM_008767709.1 | XP_008765931.1 | 7 | 1566 | |||
| XM_008767710.1 | XP_008765932.1 | 7 | 1604 | |||
| GenBank mRNA | BC166447.1 | - | 6 | 1479 | 8364461 | |

Sh3bp5l
Genomic Map