siRNA Details
| Gene Symbol: NEWGENE_1589866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750712.3 | XP_003750760.1 | 1 | 615 | ||
| Gene Symbol: Fam124b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001076121.5 | XP_001076121.3 | 1 | 615 | ||

NEWGENE_1589866
Genomic Map