siRNA Details
Gene Symbol: Fam124b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001076121.5 | XP_001076121.3 | 2 | 2085 |
Gene Symbol: NEWGENE_1589866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750712.3 | XP_003750760.1 | 2 | 2085 |
