siRNA Details
| Gene Symbol: Muc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044301.1 | NP_001037766.1 | 2 | 575 | ||
| XM_006234695.2 | XP_006234757.1 | 3 | 811 | |||
| XM_008762107.1 | XP_008760329.1 | 2 | 588 | |||
| XM_008762108.1 | XP_008760330.1 | 2 | 624 | |||
| GenBank mRNA | BC083925.1 | - | 2 | 575 | 7135360 | |

Muc15
Genomic Map