siRNA Details
| Gene Symbol: LOC683420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758873.1 | XP_008757095.1 | 4 | 2845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC683420| Gene Symbol: LOC683420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758873.1 | XP_008757095.1 | 4 | 2845 | ||