siRNA Details
| Gene Symbol: Rdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168593.1 | NP_001162065.1 | 3 | 374 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh8| Gene Symbol: Rdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168593.1 | NP_001162065.1 | 3 | 374 | ||