siRNA Details
| Gene Symbol: USP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006537.3 | NP_006528.2 | 3 | 500 | ||
| NR_046341.1 | 4 | 653 | ||||
| NR_046342.1 | 4 | 616 | ||||
| XM_017022763.1 | XP_016878252.1 | 5 | 1499 | |||
| XM_017022764.1 | XP_016878253.1 | 4 | 1435 | |||
| XM_017022765.1 | XP_016878254.1 | 4 | 641 | |||
| XM_017022766.1 | XP_016878255.1 | 5 | 684 | |||
| XM_017022767.1 | XP_016878256.1 | 6 | 1615 | |||
| XM_017022768.1 | XP_016878257.1 | 5 | 1551 | |||
| GenBank mRNA | AF073344.1 | - | 3 | 364 | ||
| AK022931.1 | - | 1 | 87 | |||
| AK300948.1 | - | 3 | 355 | |||
| AK307810.1 | - | 3 | 259 | |||
| AK309382.1 | - | 2 | 321 | |||
| AK314223.1 | - | 3 | 419 | |||
| AY461579.1 | - | 4 | 529 | |||
| BC018113.2 | - | 3 | 365 | 3916811 | ||
| BC065300.1 | - | 3 | 400 | 6146202 | ||
| BC100029.1 | - | 3 | 372 | |||
| BC107137.1 | - | 3 | 360 | 40025968 | ||
| BC107138.1 | - | 3 | 360 | 40025969 | ||
| BT007269.1 | - | 3 | 271 | |||
| BX647760.1 | - | 4 | 394 | |||
| DA557812.1 | - | 3 | 502 | |||

USP3
Genomic Map