siRNA Details
| Gene Symbol: Evl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764850.1 | XP_008763072.1 | 1 | 24 | ||
| XM_008764855.1 | XP_008763077.1 | 1 | 24 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evl| Gene Symbol: Evl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764850.1 | XP_008763072.1 | 1 | 24 | ||
| XM_008764855.1 | XP_008763077.1 | 1 | 24 | |||