siRNA Details
| Gene Symbol: USP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256702.1 | NP_001243631.1 | 9 | 1077 | ||
| NM_006537.3 | NP_006528.2 | 10 | 1209 | |||
| NR_046341.1 | 11 | 1362 | ||||
| NR_046342.1 | 11 | 1325 | ||||
| XM_017022763.1 | XP_016878252.1 | 12 | 2208 | |||
| XM_017022764.1 | XP_016878253.1 | 11 | 2144 | |||
| XM_017022765.1 | XP_016878254.1 | 11 | 1350 | |||
| XM_017022766.1 | XP_016878255.1 | 12 | 1393 | |||
| XM_017022767.1 | XP_016878256.1 | 13 | 2324 | |||
| XM_017022768.1 | XP_016878257.1 | 13 | 2398 | |||
| GenBank mRNA | AF073344.1 | - | 10 | 1076 | ||
| AF077040.1 | - | 9 | 853 | |||
| AK022931.1 | - | 8 | 796 | |||
| AK300948.1 | - | 8 | 867 | |||
| AK301236.1 | - | 9 | 993 | |||
| AK301563.1 | - | 4 | 508 | |||
| AK307810.1 | - | 10 | 968 | |||
| AK309382.1 | - | 9 | 1030 | |||
| AK314223.1 | - | 10 | 1128 | |||
| AY461579.1 | - | 11 | 1238 | |||
| BC018113.2 | - | 10 | 1074 | 3916811 | ||
| BC065300.1 | - | 10 | 1109 | 6146202 | ||
| BC100029.1 | - | 10 | 1081 | |||
| BC107137.1 | - | 10 | 1069 | 40025968 | ||
| BC107138.1 | - | 10 | 1069 | 40025969 | ||
| BT007269.1 | - | 10 | 980 | |||
| BX647760.1 | - | 11 | 1103 | |||

USP3
Genomic Map