siRNA Details
| Gene Symbol: SLC23A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005116.5 | NP_005107.4 | 4 | 569 | ||
| NM_203327.1 | NP_976072.1 | 4 | 587 | |||
| XM_011529414.2 | XP_011527716.1 | 4 | 574 | |||
| XM_011529415.2 | XP_011527717.1 | 3 | 395 | |||
| XM_011529416.2 | XP_011527718.1 | 3 | 704 | |||
| XM_011529417.2 | XP_011527719.1 | 3 | 451 | |||
| XM_017028171.1 | XP_016883660.1 | 4 | 608 | |||
| XM_017028172.1 | XP_016883661.1 | 4 | 612 | |||
| XM_017028173.1 | XP_016883662.1 | 4 | 592 | |||
| XM_017028174.1 | XP_016883663.1 | 4 | 487 | |||
| XM_017028176.1 | XP_016883665.1 | 4 | 583 | |||
| XM_017028177.1 | XP_016883666.1 | 4 | 570 | |||
| XM_017028178.1 | XP_016883667.1 | 4 | 583 | |||
| XM_017028179.1 | XP_016883668.1 | 4 | 570 | |||
| XM_017028181.1 | XP_016883670.1 | 4 | 586 | |||
| GenBank mRNA | AF058319.1 | - | 2 | 194 | ||
| AF092511.1 | - | 3 | 490 | |||
| AF164142.1 | - | 4 | 587 | |||
| AJ269478.1 | - | 2 | 194 | |||
| AJ292318.1 | - | 3 | 425 | |||
| AK296304.1 | - | 4 | 577 | |||
| AK297253.1 | - | 4 | 591 | |||
| AY380556.1 | - | 2 | 194 | |||
| BC013112.2 | - | 4 | 558 | 3923057 | ||
| D87075.2 | - | 4 | 569 | |||
| EF032501.1 | - | 2 | 194 | |||

SLC23A2
Genomic Map