siRNA Details
| Gene Symbol: Atp8b5p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238129.2 | XP_006238191.1 | 13 | 1504 | ||
| XM_008763657.1 | XP_008761879.1 | 13 | 1468 | |||
| XM_008763658.1 | XP_008761880.1 | 13 | 1504 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp8b5p| Gene Symbol: Atp8b5p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238129.2 | XP_006238191.1 | 13 | 1504 | ||
| XM_008763657.1 | XP_008761879.1 | 13 | 1468 | |||
| XM_008763658.1 | XP_008761880.1 | 13 | 1504 | |||