siRNA Details
| Gene Symbol: LOC691135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759083.1 | XP_008757305.1 | 4 | 1789 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691135| Gene Symbol: LOC691135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759083.1 | XP_008757305.1 | 4 | 1789 | ||