siRNA Details
| Gene Symbol: Nutf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007629.1 | NP_001007630.1 | 2 | 139 | ||
| XM_006255480.2 | XP_006255542.1 | 1 | 103 | |||
| GenBank mRNA | BC061569.1 | - | 2 | 139 | 6886638 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nutf2| Gene Symbol: Nutf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007629.1 | NP_001007630.1 | 2 | 139 | ||
| XM_006255480.2 | XP_006255542.1 | 1 | 103 | |||
| GenBank mRNA | BC061569.1 | - | 2 | 139 | 6886638 | |