siRNA Details
| Gene Symbol: LOC691277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109633.1 | NP_001103103.1 | 6 | 654 | ||
| XM_008767712.1 | XP_008765934.1 | 5 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691277| Gene Symbol: LOC691277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109633.1 | NP_001103103.1 | 6 | 654 | ||
| XM_008767712.1 | XP_008765934.1 | 5 | 677 | |||