siRNA Details
| Gene Symbol: LOC691286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109634.1 | NP_001103104.1 | 3 | 277 | ||
| XM_006246404.2 | XP_006246466.1 | 3 | 400 | |||
| XM_006246405.2 | XP_006246467.1 | 3 | 447 | |||
| XM_008767713.1 | XP_008765935.1 | 2 | 548 | |||

LOC691286
Genomic Map