siRNA Details
| Gene Symbol: RGD1565323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109635.1 | NP_001103105.1 | 2, 3 | 460 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565323| Gene Symbol: RGD1565323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109635.1 | NP_001103105.1 | 2, 3 | 460 | ||