siRNA Details
| Gene Symbol: Lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109637.1 | NP_001103107.1 | 5 | 641 | ||
| XM_006233231.2 | XP_006233293.1 | 5 | 628 | |||
| GenBank mRNA | BC158606.1 | - | 5 | 597 | ||
| FQ215425.1 | - | 5 | 582 | |||

Lrrc39
Genomic Map