siRNA Details
| Gene Symbol: LOC687345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765194.1 | XP_008763416.1 | 4 | 1177 | ||
| Gene Symbol: Cyp4f37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271352.1 | NP_001258281.1 | 9 | 1427 | ||

LOC687345
Genomic Map