siRNA Details
| Gene Symbol: Draxin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239423.2 | XP_006239485.1 | 2 | 278 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Draxin| Gene Symbol: Draxin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239423.2 | XP_006239485.1 | 2 | 278 | ||