siRNA Details
| Gene Symbol: LOC691325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001077719.3 | XP_001077719.1 | 2 | 307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691325| Gene Symbol: LOC691325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001077719.3 | XP_001077719.1 | 2 | 307 | ||