siRNA Details
| Gene Symbol: Hus1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134846.1 | NP_001128318.1 | 1 | 221 | ||
| Gene Symbol: LOC103690165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771605.1 | XP_008769827.1 | 1 | 2717 | ||

Hus1b
Genomic Map