siRNA Details
| Gene Symbol: LOC100911196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228306.2 | XP_006228368.1 | 1 | 2146 | ||
| XM_008759010.1 | XP_008757232.1 | 1 | 2677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100911196| Gene Symbol: LOC100911196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228306.2 | XP_006228368.1 | 1 | 2146 | ||
| XM_008759010.1 | XP_008757232.1 | 1 | 2677 | |||