siRNA Details
| Gene Symbol: Tmem190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001078306.4 | XP_001078306.2 | 2 | 1496 | ||
| XM_008759018.1 | XP_008757240.1 | 2 | 1496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem190| Gene Symbol: Tmem190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001078306.4 | XP_001078306.2 | 2 | 1496 | ||
| XM_008759018.1 | XP_008757240.1 | 2 | 1496 | |||