siRNA Details
| Gene Symbol: Cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108982.2 | NP_001102452.2 | 4 | 558 | ||
| XM_006240104.2 | XP_006240166.1 | 4 | 645 | |||
| GenBank mRNA | FQ215859.1 | - | 3 | 294 | ||
| FQ224178.1 | - | 4 | 234 | |||
| FQ228820.1 | - | 4 | 561 | |||
| FQ228857.1 | - | 4 | 565 | |||

Cfl2
Genomic Map