siRNA Details
| Gene Symbol: Prl5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044702.1 | NP_001038167.1 | 2 | 200 | ||
| GenBank mRNA | DQ329280.1 | - | 2 | 200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl5a2| Gene Symbol: Prl5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044702.1 | NP_001038167.1 | 2 | 200 | ||
| GenBank mRNA | DQ329280.1 | - | 2 | 200 | ||