siRNA Details
| Gene Symbol: Cpne6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191113.1 | NP_001178042.1 | 5, 6 | 600 | ||
| GenBank mRNA | AM746509.1 | - | 4, 5 | 487 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpne6| Gene Symbol: Cpne6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191113.1 | NP_001178042.1 | 5, 6 | 600 | ||
| GenBank mRNA | AM746509.1 | - | 4, 5 | 487 | ||