siRNA Details
| Gene Symbol: Katnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109645.2 | NP_001103115.1 | 6 | 689 | ||
| XM_006234712.2 | XP_006234774.1 | 6 | 742 | |||
| GenBank mRNA | BC161951.1 | - | 6 | 689 | 9026012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Katnbl1| Gene Symbol: Katnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109645.2 | NP_001103115.1 | 6 | 689 | ||
| XM_006234712.2 | XP_006234774.1 | 6 | 742 | |||
| GenBank mRNA | BC161951.1 | - | 6 | 689 | 9026012 | |