siRNA Details
| Gene Symbol: DOPEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320714.1 | NP_001307643.1 | 19 | 4231 | ||
| NM_005128.3 | NP_005119.2 | 19 | 4122 | |||
| XM_017028509.1 | XP_016883998.1 | 16 | 3625 | |||
| GenBank mRNA | AB023150.2 | - | 16 | 3591 | ||
| AJ237839.2 | - | 19 | 4128 | |||

DOPEY2
Genomic Map