siRNA Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135562.1 | NP_001129034.1 | 3, 4 | 300 | ||
| XM_008764572.1 | XP_008762794.1 | 3, 4 | 300 | |||
| GenBank mRNA | BC158763.1 | - | 3, 4 | 300 | 7109165 | |
| BC161990.1 | - | 3, 4 | 246 | 9032684 | ||
| FQ215147.1 | - | 3, 4 | 238 | |||

Acp1
Genomic Map