siRNA Details
| Gene Symbol: Gzmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191116.1 | NP_001178045.1 | 5 | 661 | ||
| XM_008770701.1 | XP_008768923.1 | 6 | 759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gzmn| Gene Symbol: Gzmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191116.1 | NP_001178045.1 | 5 | 661 | ||
| XM_008770701.1 | XP_008768923.1 | 6 | 759 | |||