siRNA Details
| Gene Symbol: REC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048205.1 | NP_001041670.1 | 3 | 770 | ||
| NM_005132.2 | NP_005123.2 | 4 | 669 | |||
| XM_011537421.1 | XP_011535723.1 | 3 | 942 | |||
| XM_017021841.1 | XP_016877330.1 | 3 | 739 | |||
| GenBank mRNA | AF006264.1 | - | 3 | 242 | ||
| AK001435.1 | - | 4 | 669 | |||
| AK291191.1 | - | 4 | 801 | |||
| AL832973.1 | - | 3 | 660 | |||
| BC004159.2 | - | 3 | 326 | 2822680 | ||
| BC010887.1 | - | 3 | 703 | 3535425 | ||
| CR457263.1 | - | 3 | 242 | |||
| DB294585.1 | - | 3 | 764 | |||

REC8
Genomic Map