siRNA Details
| Gene Symbol: Lpar6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045843.1 | NP_001039308.1 | 1 | 1166 | ||
| GenBank mRNA | BC098703.1 | - | 1 | 1166 | 7389453 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpar6| Gene Symbol: Lpar6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045843.1 | NP_001039308.1 | 1 | 1166 | ||
| GenBank mRNA | BC098703.1 | - | 1 | 1166 | 7389453 | |