siRNA Details
| Gene Symbol: LOC691849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198796.1 | NP_001185725.1 | 2 | 384 | ||
| GenBank mRNA | FM128707.1 | - | 2 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691849| Gene Symbol: LOC691849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198796.1 | NP_001185725.1 | 2 | 384 | ||
| GenBank mRNA | FM128707.1 | - | 2 | 378 | ||