siRNA Details
| Gene Symbol: LOC102554917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228360.1 | XP_006228422.1 | 4 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102554917| Gene Symbol: LOC102554917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228360.1 | XP_006228422.1 | 4 | 372 | ||