siRNA Details
| Gene Symbol: LOC691909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134694.2 | NP_001128166.2 | 5 | 951 | ||
| GenBank mRNA | BC158625.1 | - | 5 | 951 | 7454926 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691909| Gene Symbol: LOC691909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134694.2 | NP_001128166.2 | 5 | 951 | ||
| GenBank mRNA | BC158625.1 | - | 5 | 951 | 7454926 | |